Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Dodavatel: Thermo Fisher Scientific
12386.FF
12386.FI
12386.FFEA
4740
CZK
12386.FF
12386.FI
Erbium ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.25 mm (0.01 in)
Erbium
Purity: 99.9%
CAS No.: 7440-52-0
Physical Form: ≈1.4g/25x25mm
Odour: Odourless
MDL No.: MFCD00010987
Vzorec:
Er Storage Temperature: Ambient |
MDL Number:
MFCD00010987 CAS číslo: 7440-52-0 EINECS: 231-160-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...