Erbium ≥99.9% (REO, rare earth oxide basis), pieces
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Packaged under argon.
00111.09
00111.18
00111.09EA
5220
CZK
00111.09
00111.18
Erbium ≥99.9% (REO, rare earth oxide basis), pieces
Erbium
Purity: 99.9%
CAS No.: 7440-52-0
Physical Form: Packaged under argon in glass bottle
Appearance: Silvery grey
UN No.: 3178
MDL No.: MFCD00010987
Vzorec:
Er MW: 167,26 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010987 CAS číslo: 7440-52-0 EINECS: 231-160-1 UN: 3178 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...