Cer ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
44037.DJ
44037.DM
44037.HF
44037.DJEA
13110
CZK
44037.DJ
44037.DM
44037.HF
Cer ≥99.9% (metals basis excluding Ta), rods, Ø 6.35 mm (0.25 in)
Cerium
Purity: 99.9%
CAS No.: 7440-45-1
Molecular Formula: Packaged in mylar under argon
Physical Form: ≈0.21g/mm, Cast rod, diameter is approximate
Appearance: Silver
Odour: Odourless
UN No.: 1333
MDL No.: MFCD00010924
Vzorec:
Ce MW: 140,12 g/mol Bod varu: 3468 °C (1013 hPa) Bod tání: 795 °C Hustota: ∼6,8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010924 CAS číslo: 7440-45-1 EINECS: 231-154-9 UN: 1333 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...