Cer ≥99% (REO, rare earth oxide basis), ingot
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
43977.30
43977.A1
43977.30EA
6870
CZK
43977.30
43977.A1
Cer ≥99% (REO, rare earth oxide basis), ingot
Cerium
Purity: 99%
CAS No.: 7440-45-1
Physical Form: Packaged under mineral oil
Appearance: Gray
Odour: Odourless
UN No.: 1333
MDL No.: MFCD00010924
Vzorec:
Ce Bod varu: 3468 °C (1013 hPa) Bod tání: 795 °C Hustota: ∼6,8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010924 CAS číslo: 7440-45-1 EINECS: 231-154-9 UN: 1333 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...