Tisk…

Cer ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

10141.FF 10141.FI
10141.FFEA 7280 CZK
10141.FF 10141.FI
Cer ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.1 mm (0.004 in)
Cerium

Purity: 99.9% CAS No.: 7440-45-1 Physical Form: ≈0.42g/25x25mm, Packaged in mylar under argon Appearance: Silver-grey Odour: Odourless UN No.: 1333 MDL No.: MFCD00010924

Vzorec: Ce
Bod varu: 3468 °C (1013 hPa)
Bod tání: 795 °C
Hustota: ∼6,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010924
CAS číslo: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR