Cer ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Dodavatel: Thermo Scientific
|
Nebezpečí
|
00591.FF
00591.FI
00591.FFEA
6730
CZK
00591.FF
00591.FI
Cer ≥99.9% (REO, rare earth oxide basis), foil, Thickness: 0.62 mm (0.024 in)
Cerium
Purity: 99.9%
CAS No.: 7440-45-1
Physical Form: ≈2.6g/25x25mm, Packaged in mylar under argon
Appearance: Silver-grey
Odour: Odourless
UN No.: 1333
MDL No.: MFCD00010924
Vzorec:
Ce MW: 140,12 g/mol Bod varu: 3468 °C (1013 hPa) Bod tání: 795 °C Hustota: ∼6,8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010924 CAS číslo: 7440-45-1 EINECS: 231-154-9 UN: 1333 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...