Tisk…

Telur, powder, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

266418-100G 266418-25G 266418-500G
SIAL266418-100GEA 7080 CZK
SIAL266418-100G SIAL266418-25G SIAL266418-500G
Telur, powder, Sigma-Aldrich®
Telur
Vzorec: Te
MW: 127.6
Bod varu: 990
Bod tání: 450
MDL Number: MFCD00134062
CAS číslo: 13494-80-9
EINECS: 236-813-4
Merck Index: 12,09273

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR