křemík ≥99.999% (metals basis), plate, Thickness: 9.5 mm (0.37 in)
Dodavatel: Thermo Fisher Scientific
Varování
|
39110.FF
39110.FI
39110.FFEA
4400
CZK
39110.FF
39110.FI
křemík ≥99.999% (metals basis), plate, Thickness: 9.5 mm (0.37 in)
křemík
Purity: 99.999%
CAS No.: 7440-21-3
Physical Form: ≈14.32g/25x25mm
Odour: Odourless
MDL No.: MFCD00085311
Vzorec:
Si MW: 28.09 g/mol Hustota: 2.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS číslo: 7440-21-3 EINECS: 231-130-8 Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...