křemík ≥99.999% (metals basis), granular virgin poly fines
Dodavatel: Thermo Fisher Scientific
|
Varování
|
38542.14
38542.22
38542.14EA
1530
CZK
38542.14
38542.22
křemík ≥99.999% (metals basis), granular virgin poly fines
křemík
Purity: 99.999%
CAS No.: 7440-21-3
Physical Form: ~1/4in and down (fm chunks to powder)
Odour: Odourless
MDL No.: MFCD00085311
Vzorec:
Si MW: 28.09 g/mol Hustota: 2.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS číslo: 7440-21-3 EINECS: 231-130-8 Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...