Tisk…

křemík ≥99.999% (metals basis), crystalline powder -100+325 mesh

Dodavatel: Thermo Fisher Scientific
Varování

36212.09 36212.18 36212.30
36212.09EA 2550 CZK
36212.09 36212.18 36212.30
křemík ≥99.999% (metals basis), crystalline powder -100+325 mesh
křemík

Purity: 99.999% CAS No.: 7440-21-3 Appearance: White Odour: Odourless UN No.: 1346 MDL No.: MFCD00085311

Vzorec: Si
MW: 28.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS číslo: 7440-21-3
EINECS: 231-130-8
UN: 1346
ADR: 4.1 III
Merck Index: 12,08635

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR