Tisk…
Grafit 99,8 Metals basis, foil, Thickness: 0.254 mm (0.01 in)
Dodavatel: Thermo Fisher Scientific
10832.CG 10832.VA 10832.VU
10832.CGEA 3610 CZK
10832.CG 10832.VA 10832.VU
Grafit 99,8 Metals basis, foil, Thickness: 0.254 mm (0.01 in)
Grafit
Purity: 99.8% Density: 1.12 MDL No.: MFCD00144065
Vzorec: C
MW: 12.01
Bod tání: 3827
Hustota: 1.12
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS číslo: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560