Tisk…

Glassy carbon, powder 200-400 µm Type 1

Dodavatel: Thermo Fisher Scientific
Varování

42130.09 42130.18
42130.09EA 2360 CZK
42130.09 42130.18
Glassy carbon, powder 200-400 µm Type 1
Glassy carbon
Vzorec: C
MW: 12.01
Bod tání: 3652
Hustota: 1.95
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS číslo: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1 III
Merck Index: 13,01818

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR