Glassy carbon, spherical powder 630-1000 µm
Dodavatel: Thermo Scientific
|
Varování
|
41530.18
41530.30
41530.18EA
6180
CZK
41530.18
41530.30
Glassy carbon, spherical powder 630-1000 µm
Glassy carbon
Vzorec:
C MW: 12,01 g/mol Bod tání: 3652 °C Hustota: 1,95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS číslo: 7440-44-0 EINECS: 231-153-3 UN: 1325 ADR: 4.1,III Merck Index: 13,01818 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...