Tisk…

Germanium, powder, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

203351-50G 203351-10G
SIAL203351-50GEA 30930 CZK
SIAL203351-50G SIAL203351-10G
Germanium, powder, Sigma-Aldrich®
Germanium
Vzorec: Ge
MW: 72,61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS číslo: 7440-56-4
EINECS: 231-164-3
UN: 3089
ADR: 4.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR