Germanium, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
263230-10G
263230-50G
SIAL263230-10GEA
12250
CZK
SIAL263230-10G
SIAL263230-50G
Germanium, Sigma-Aldrich®
Germanium
Vzorec:
Ge MW: 72,61 g/mol Bod varu: 2830 °C (1013 hPa) Bod tání: 937,2 °C Hustota: 5,323 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085310 CAS číslo: 7440-56-4 EINECS: 231-164-3 Merck Index: 12,04415 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...