Tisk…

Germanium ≥99.9999% (metals basis), pieces ≤ 2 cm (0.8 in)

Dodavatel: Thermo Scientific

00123.09 00123.18
00123.09EA 6280 CZK
00123.09 00123.18
Germanium ≥99.9999% (metals basis), pieces ≤ 2 cm (0.8 in)
Germanium

Purity: 99.9999% CAS No.: 7440-56-4 Physical Form: Electrical Resistivity: 40 ohm-cm Odour: Odourless MDL No.: MFCD00085310

Vzorec: Ge
MW: 72,61 g/mol
Bod varu: 2830 °C (1013 hPa)
Bod tání: 937,2 °C
Hustota: 5,323 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085310
CAS číslo: 7440-56-4
EINECS: 231-164-3
Merck Index: 12,04415

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR