Indium ≥99.999% (metals basis), ingot
Dodavatel: Thermo Scientific
14720.09
14720.18
14720.09EA
4810
CZK
14720.09
14720.18
Indium ≥99.999% (metals basis), ingot
Indium
Purity: 99.999%
CAS No.: 7440-74-6
Physical Form: Soft white ingot
Odour: Odourless
MDL No.: MFCD00134048
Vzorec:
In MW: 114,82 g/mol Bod varu: 2000 °C (1013 hPa) Bod tání: 156 °C Hustota: 7,3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS číslo: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...