Tisk…

Indium ≥99.99% (metals basis), foil, Thickness: 0.25 mm (0.01 in)

Dodavatel: Thermo Fisher Scientific

13982.FI 13982.GJ 13982.HW
13982.FIEA 4710 CZK
13982.FI 13982.GJ 13982.HW
Indium ≥99.99% (metals basis), foil, Thickness: 0.25 mm (0.01 in)
Indium

Purity: 99.99% CAS No.: 7440-74-6 Physical Form: ≈4.6g/50x50mm Odour: Odourless MDL No.: MFCD00134048

Vzorec: In
Bod varu: 2000 °C (1013 hPa)
Bod tání: 156 °C
Hustota: 7,3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS číslo: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR