Tisk…
Indium ≥99.99% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Dodavatel: Thermo Fisher Scientific
12206.FI 12206.FY 12206.GJ 12206.HW
12206.FIEA 3290 CZK
12206.FI 12206.FY 12206.GJ 12206.HW
Indium ≥99.99% (metals basis), foil, Thickness: 0.127 mm (0.005 in)
Indium
Purity: 99.99% CAS No.: 7440-74-6 Physical Form: ≈2.3g/50x50mm Odour: Odourless MDL No.: MFCD00134048
Vzorec: In
MW: 114.82
Bod varu: 2000
Bod tání: 156
Hustota: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS číslo: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971