hořčík ≥99.8%, powder -325 mesh
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
10233.09
10233.22
10233.36
10233.57
10233.09EA
2410
CZK
10233.09
10233.22
10233.36
10233.57
hořčík ≥99.8%, powder -325 mesh
hořčík
Purity: 99.8%
CAS No.: 7439-95-4
Appearance: Silver-grey
Odour: Odourless
UN No.: 1418
MDL No.: MFCD00085308
Vzorec:
Mg Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS číslo: 7439-95-4 EINECS: 231-104-6 UN: 1418 ADR: 4.3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...