Tisk…

hořčík ≥99.9% (metals basis), foil, hard, Thickness: 1.0 mm (0.04 in)

Dodavatel: Thermo Fisher Scientific
Nebezpečí

40604.FF 40604.FI 40604.GH
40604.FFEA 7250 CZK
40604.FF 40604.FI 40604.GH
hořčík ≥99.9% (metals basis), foil, hard, Thickness: 1.0 mm (0.04 in)
hořčík

Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈1.1g/25x25mm, Temper: as rolled Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308

Vzorec: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS číslo: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR