Barium, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
441880-25G
441880-5G
SIAL441880-25GEA
51050
CZK
SIAL441880-25G
SIAL441880-5G
Barium, Sigma-Aldrich®
Barium
Vzorec:
Ba Bod varu: 1640 °C (1013 hPa) Bod tání: 725 °C Hustota: 3,51 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134031 CAS číslo: 7440-39-3 EINECS: 231-149-1 UN: 1400 ADR: 4.3,II Merck Index: 12,00991 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...