Rubidium, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
276332-1G
276332-5G
SIAL276332-1GEA
7740
CZK
SIAL276332-1G
SIAL276332-5G
Rubidium, Sigma-Aldrich®
Rubidium
Vzorec:
Rb Bod varu: 700 °C (1013 hPa) Bod tání: 39 °C Hustota: 0,83 g/cm³ (1399.85 °C) |
MDL Number:
MFCD00134055 CAS číslo: 7440-17-7 EINECS: 231-126-6 UN: 1423 ADR: 4.3,I Merck Index: 12,08439 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...