Tisk…

Dulcoside B ≥98%, crystalline

Dodavatel: Cayman Chemical

Synonyms: Rebaudioside C, (4R,4aS,6aR,9S,11aR,11bS)-(2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl 9-(((2S,3R,4S,5R,6R)-5-hydroxy-6-(hydroxymethyl)-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-4,11b-dimethyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylate

11895-1 11895-10 11895-25 11895-5
CAYM11895-1EA 1320 CZK
CAYM11895-1 CAYM11895-10 CAYM11895-25 CAYM11895-5
Dulcoside B ≥98%, crystalline
Dulcoside B

Rebaudioside C is a natural non-caloric sweetener.


It is one of the predominant steviol glycosides isolated from S. rebaudiana leaves. Rebaudioside C, similarly to rebaudioside A is metabolised to steviol a compound whose safety is widely studied.


Soluble in organic solvents such as DMSO and dimethyl formamide purged with inert gas. Its solubility is approximately 1 mg/ml.

Vzorec: C₄₄H₇₀O₂₂
MW: 951,01 g/mol
Storage Temperature: Low Temperature Freezer (–40°C)
MDL Number: MFCD02752304
CAS číslo: 63550-99-2

Objednat

Výsledky specifikačního testu

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR