Tisk…
GC columns for blood alcohol concentration (BAC)
Dodavatel: Agilent
123-9334UI 123-9434UI 125-9434UI 125-9334UI
HEWL123-9334UIEA 29740 CZK
HEWL123-9334UI HEWL123-9434UI HEWL125-9434UI HEWL125-9334UI
GC columns for blood alcohol concentration (BAC)
Kolony chromatografické
These columns are suitable for the analysis of Blood Alcohol Concentration (BAC) using Headspace GC dual FID or headspace GC/MS.