Tisk…
Racks for centrifuge tube cryoboxes
Dodavatel: TENAK
TE25208 TE25206 TE25204 TE25202 TE25200
479-0849EA 6640 CZK
479-0849 479-0848 479-0847 479-0846 479-0845
Racks for centrifuge tube cryoboxes
Stojany Poličky pro kryoboxy
Stainless steel, sliding shelf.
  • Sliding shelf rack design
  • Can be used with all freezer brands
  • For boxes 130 mm height
For upright freezers. The racks are specially designed for use with boxes for centrifuge tubes. Boxes with exterior dimensions up to H×W 132×137 mm, the length can in principle be as long as the shelf.
Informace pro objednávání: Delivered without cryoboxes