Tisk…
Protease Inhibitor I aldehyde
Dodavatel: ENZO LIFE SCIENCES
ALX260089M001 ALX260089M005
ENZOALX260089M001EA 5710 CZK
ENZOALX260089M001 ENZOALX260089M005
Protease Inhibitor I aldehyde
Proteiny
Cell permeable inhibitor of the 26S proteasome (MCP; multicatalytic proteinase complex) in HT4 cells. Causes accumulation of ubiquitinylated proteins in neuronal cells. Prevents the activation of NF-κB in response to TNF-α or okadaic acid through inhibition of IκBα degradation.
White powder.
Soluble in 100% ethanol (30 mg/ml), DMSO (30 mg/ml) or methanol (50 mg/ml).