Tisk…
Human recombinant Trap1,HIS Tag
Dodavatel: ENZO LIFE SCIENCES
ADISPP848050 ADISPP848200
ENZOADISPP848050EA 8670 CZK
ENZOADISPP848050 ENZOADISPP848200
Human recombinant Trap1,HIS Tag
Proteiny
Produced in E. coli. Human TRAP1 (aa 60-704) is fused at the N-terminus to a His-tag.