Tisk…
Anti-IgE Mouse Monoclonal Antibody [clone: I27]
Dodavatel: Axxora
ENZOENZABS2270100EA 15370 CZK
ALEXENZABS2270100 ENZOENZABS2270100
Anti-IgE Mouse Monoclonal Antibody [clone: I27]
Protilátky
Recommended Applications: Flow Cytometry, WB, FUNC
Species reactivity: Human
Type: Secondary
Antigen: IgE
Clonality: Monoclonal
Clone: I27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human