Tisk…

Anti-HA-tag Rabbit Polyclonal Antibody

Dodavatel: ENZO LIFE SCIENCES
ENZABS2030200 ENZABS2030500
ENZOENZABS2030200EA 20900 CZK
ENZOENZABS2030200 ENZOENZABS2030500
Anti-HA-tag Rabbit Polyclonal Antibody
Protilátky
Recommended Applications: IP, WB

Species reactivity:

Type: Primary
Antigen: HA-tag
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR