Tisk…
Anti-HA-tag Mouse Monoclonal Antibody [clone: 16B12]
Dodavatel: ENZO LIFE SCIENCES
ENZABS1180200 ENZABS1180500 ENZABS1181000
ENZOENZABS1180200EA 14060 CZK
ENZOENZABS1180200 ENZOENZABS1180500 ENZOENZABS1181000
Anti-HA-tag Mouse Monoclonal Antibody [clone: 16B12]
Protilátky
Recommended Applications: IF, IHC, IP, WB
Species reactivity:
Type: Primary
Antigen: HA-tag
Clonality: Monoclonal
Clone: 16B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse