Tisk…
Anti-HSP90A Rabbit Polyclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADISPS771PE050 ADISPS771PE200
ENZOADISPS771PE050EA 8730 CZK
ENZOADISPS771PE050 ENZOADISPS771PE200
Anti-HSP90A Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: HSP90A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Pig, Monkey, Hamster, Guinea pig, Fish, Dog, Bovine, Beluga