Tisk…
Anti-GRP78 Rabbit Polyclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADISPA7690050 ADISPA7690200
ENZOADISPA7690050EA 5190 CZK
ENZOADISPA7690050 ENZOADISPA7690200
Anti-GRP78 Rabbit Polyclonal Antibody
Protilátky
Type: Primary
Antigen: Grp78
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Xenopus, Rabbit, Monkey, Hamster, Fungus, Bovine