Tisk…

Anti-ERK1 + ERK2 Rabbit Polyclonal Antibody

Dodavatel: ENZO LIFE SCIENCES
ADIKAPMA001D ADIKAPMA001F
ENZOADIKAPMA001DEA 6990 CZK
ENZOADIKAPMA001D ENZOADIKAPMA001F
Anti-ERK1 + ERK2 Rabbit Polyclonal Antibody
Protilátky
The Extracellular regulated kinases, Erk-1 and Erk-2, both play a critical role in cellular proliferation. These extracellular kinases, are known to reside downstream of the Mitogen activated protein kinase/Extracellular protein kinases referred to as MEK. Both MEK-1 and MEK-2 phosphorylate and activate Erk on the conserved TEY motif within the activation loop of the kinase.

Type: Primary
Antigen: ERK1 + ERK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Xenopus, Sheep, Drosophila, Chicken, Bovine
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR