Tisk…

Anti-LTC4 Mouse Monoclonal Antibody [clone: 6E7]

Dodavatel: ENZO LIFE SCIENCES
ADI9059020050 ADI9059020200
ENZOADI9059020050EA 8700 CZK
ENZOADI9059020050 ENZOADI9059020200
Anti-LTC4 Mouse Monoclonal Antibody [clone: 6E7]
Protilátky
Leukotrienes are metabolic products of arachidonic acid formed by the action of 5-lipoxygenase (5-LO), with the principal products being leukotrienes C4 (LTC4) and B4 (LTB4). Leukotriene B4 (LTB4) stimulates leukocyte functions including lysosomal enzyme release, adhesion, and aggregation of polymorphonuclear leukocytes. LTB4 has been implicated as a potent mediator of inflammatory diseases and immunoregulation. The cysteinyl leukotrienes (LTC4, LTD4, and LTE4) are formed by addition of cysteine derivatives to LTA4. The conversion of LTA4 to LTC4 by the enzyme LTC4 synthase limits the rate of cysteinyl leukotriene formation. Cysteinyl leukotrienes are potent lipid mediators in inflammation, inducing smooth muscle contractions and increased capillary permeability.

Type: Primary
Antigen: LTC4
Clonality: Monoclonal
Clone: 6E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR