Tisk…
Anti-SP100 Rabbit Polyclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
BMLPW03250025 BMLPW03250100
ENZOBMLPW03250025EA 3680 CZK
ENZOBMLPW03250025 ENZOBMLPW03250100
Anti-SP100 Rabbit Polyclonal Antibody
Protilátky
Sp100 is an interferon-inducible protein and a major constituent of distinct nuclear domains, known as nuclear dots (NDs) or PML bodies. NDs play a role in autoimmunity, virus infections, and in the etiology of acute promyelocytic leukaemia. SUMO modification of Sp100 has been shown to promote interaction of Sp100 with non-histone chromosomal proteins, suggesting a regulatory role in chromatin organization and in functional interplay between these nuclear bodies and chromatin.
  • For Immunocytochemistry, Western Blot
Recognizes human Sp100 and SUMO modified Sp100 species.
Type: Primary
Antigen: SP100
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human