Tisk…
Anti-CTNNA1 Mouse Monoclonal Antibody
Dodavatel: ENZO LIFE SCIENCES AG
ENZOALX804101C100EA 17470 CZK
ALEXALX804101C100 ENZOALX804101C100
Anti-CTNNA1 Mouse Monoclonal Antibody
Protilátky
Host: Mouse, Isotype: IgG1
  • For Immunocytochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat and dog alpha-catenin. Does not cross-react with beta-catenins.
Reconstitute with 1ml distilled water (15 min., RT).
Type: Primary
Antigen: CTNNA1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Objednat