Tisk…

Anti-Aldosterone Receptor Mouse Monoclonal Antibody

Dodavatel: Enzo Life Sciences
ENZOALX804099R100EA 24520 CZK
ALEXALX804099R100 ENZOALX804099R100
Anti-Aldosterone Receptor Mouse Monoclonal Antibody
Protilátky
Host: Mouse, Isotype: IgG1

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human, rat, rabbit and chicken MR (mineralocorticoid receptor). Detects a band of ~116kDa by Western blot.

Type: Primary
Antigen: Aldosterone Receptor
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR