Tisk…
Anti-HSP90 Mouse Monoclonal Antibody
Dodavatel: ENZO LIFE SCIENCES AG
ENZOALX804079R400EA 17440 CZK
ALEXALX804079R400 ENZOALX804079R400
Anti-HSP90 Mouse Monoclonal Antibody
Protilátky
Host: Mouse, Isotype: IgM
  • For Immunoprecipitation
Recognizes human, mouse, rat, rainbow trout and chicken HSP90.
Type: Primary
Antigen: HSP90
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human