Tisk…
Anti-Transferrin Rabbit Polyclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADIVAPEN004D ADIVAPEN004F
ENZOADIVAPEN004DEA 6870 CZK
ENZOADIVAPEN004D ENZOADIVAPEN004F
Anti-Transferrin Rabbit Polyclonal Antibody
Protilátky
Transferrin is a serum glycoprotein involved in iron transport. Transferrin exists in two forms, the iron-free Apotransferrin, which binds two Fe3+ ions to generate Ferrotransferrin. Apotransferrin remains tightly bound to its receptor until the complex is recycled to the cell surface, where Apotransferrin is released into the extracellular space to recruit more Fe3+ ions.
  • For Immunoprecipitation, Western Blot
Recognizes human, mouse, rat, dog and guinea pig Transferrin. Detects a band of ~76kDa by Western blot.
Type: Primary
Antigen: TRANSFERRIN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human