Tisk…
Anti-ESR1 Mouse Monoclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADISRA1010D ADISRA1010F
ENZOADISRA1010DEA 7840 CZK
ENZOADISRA1010D ENZOADISRA1010F
Anti-ESR1 Mouse Monoclonal Antibody
Protilátky
The Estrogen receptors (ER), mainly ERalpha and ERbeta, are members of the nuclear steroid hormone receptor family and are composed of three domains, an N-terminal domain required for maximal transactivation function (TAF), a DNA binding domain, and a steroid binding domain. Estrogen has pleiotropic effects on the growth, development, and maintenance of diverse tissues.
  • For Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Recognizes human, mouse, rat and hamster estrogen receptor alpha. Detects a band of 66.3kDa by WesternMore Product Information