Tisk…
Anti-HSP25 Rabbit Polyclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADISPA803D ADISPA803F
ENZOADISPA803DEA 6870 CZK
ENZOADISPA803D ENZOADISPA803F
Anti-HSP25 Rabbit Polyclonal Antibody
Protilátky
Hsp27 is one of the most common members of the highly conserved and ubiquitously expressed family of small heat shock proteins (sHsp), which also includes alphaB-crystallin. It is characterized by a conserved C-terminal alpha-crystallin domain consisting of two anti-parallel beta-sheets that promote oligomer formation required for its primary chaperone function as inhibitor of irreversible protein aggregation. Hsp27 oligomerization is modulated by post-translational phosphorylation of Hsp27 at three serine residues, Ser15, Ser78, and Ser82, by a variety of protein kinases including MAPKAPK-3,More Product Information
New Product
Objednat
Produktová specifikace
Symbol antigenu HSP25
Název antigenu Heat Shock Protein 25
Environmentally Preferable
Synonyma antigenu Stress-responsive protein 27|Heat shock 27 kDa protein|SRP27|HSP27|HspB1|HSP25|Estrogen-regulated 24 kDa protein|HSPB1|HSP 27|Heat shock protein beta-1|28 kDa heat shock protein
Druh protilátky Primární
Klonovalita Polyclonal
Reaktivita Pig,Monkey,Human
Hostitel Rabbit
Izotyp
Western blot Yes
ELISA Yes
Imunochemie Yes
Imunoprecipitace Yes
Formulář Liquid. In PBS containing 50% glycerol and 0,09% sodium azide.
Čištění Protein A affinity purified