Tisk…

Anti-HSP104 Rabbit Polyclonal Antibody

Dodavatel: ENZO LIFE SCIENCES
ADISPA1040D ADISPA1040F
ENZOADISPA1040DEA 6870 CZK
ENZOADISPA1040D ENZOADISPA1040F
Anti-HSP104 Rabbit Polyclonal Antibody
Protilátky
The 104 kDa yeast heat shock protein (Hsp104) is a cytosolic member of the Hsp100 family of proteins. Hsp104 cooperates with Hsp40 and Hsp70 co-chaperones in yeast in the reactivation of heat-damaged proteins. Hsp104 is also critical for the establishment and maintenance of the [PSI] prion phenotype in Saccharomyces cerevesiae.

  • For Western Blot

Recognizes yeast HSP104. Detects a band of ~104kDa by Western blot.

Type: Primary
Antigen: HSP104
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR