Tisk…
Anti-SOD2 Rabbit Polyclonal Antibody
Dodavatel: ENZO LIFE SCIENCES
ADISOD110D ADISOD110F
ENZOADISOD110DEA 7840 CZK
ENZOADISOD110D ENZOADISOD110F
Anti-SOD2 Rabbit Polyclonal Antibody
Protilátky
SOD (Superoxide dismutase) is responsible for the elimination of cytotoxic active oxygen by catalyzing the dismutation of the superoxide radical to oxygen and hydrogen peroxide. There are three SOD isoenzymes in mammalian cells, they are: EC SOD (extracellular SOD), Cu/Zn SOD (copper and zinc-containing SOD) and Mn SOD (manganese-containing SOD).
  • For Immunohistochemistry, Western Blot
Recognizes human, mouse, rat, bovine, dog, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus Mn SOD. Detects a band of ~25kDa by Western blot.
Type: Primary
Antigen: SOD2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human