Tisk…

Anti-IgG1 Mouse Monoclonal Antibody (PE (Phycoerythrin))

Dodavatel: ENZO LIFE SCIENCES
ADISAB600PED ADISAB600PEF
ENZOADISAB600PEDEA 8270 CZK
ENZOADISAB600PED ENZOADISAB600PEF
Anti-IgG1 Mouse Monoclonal Antibody (PE (Phycoerythrin))
Protilátky
Isotype matched controls are useful in flow cytometry to determine a gross estimate of non-specific binding that may be observed with a specific antibody of interest. In general, an isotype control should be of the same species and isotype as the antibody of interest, used at the same concentration, and conjugated to the same fluorophore. This reagent is an IgG1 isotype mouse monoclonal antibody (Clone # MOPC-21) conjugated to phycoerythrin (PE). It is intended for use as an isotype control antibody for flow cytometry.

Type: Primary
Antigen: IgG1
Clonality: Monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: pan
Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR