2-Carboxyethyl acrylate inhibited, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ) 900 – 1100 ppm
M-Clarity™ quality level = MQ200
2-Carboxyethyl acrylate are more reactive in its salt form than acylic acid, allowing high levels of incorporation over awide pH range.
2-Carboxyethyl acrylate can be polymerized in solution or emulsion to produce acrylic, vinyl-acrylic, or styrenic-acrylic polymers with greater flexibility due to lower glass transition of itshomopolymer (<30C) and improved adhesion.
Vzorec:
C₆H₈o₄ Bod varu: 103 °C (19 mmHg) Hustota: 1,214 g/ml (25 °C) (l) Bod vzplanutí: 110 °C |
MDL Number:
MFCD00040709 CAS číslo: 24615-84-7 UN: 3625 ADR: ,II |
Výsledky specifikačního testu
Colour | Colourless |
Form | Liquid |
Infrared spectrum | Conforms to structure |
Proton NMR spectra | Conforms to structure |
MEHQ as inhibitor | 900 – 1100 ppm |
Refractive index n20/D | 1,457 (l) |
pH (10 wt.% in H₂O) | 2,95 |
Solubility (H₂O) | Soluble |
Density (25 °C) | 1,214 g/ml |
Storage temperature | 2 – 8 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...