trans-2-Chloromethylvinylboronic acid ≥95%, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
556599-1G
556599-5G
SIAL556599-1GEA
2680
CZK
SIAL556599-1G
SIAL556599-5G
trans-2-Chloromethylvinylboronic acid ≥95%, Sigma-Aldrich®
trans-2-Chloromethylvinylboronic acid
- Contains varying amounts of anhydride
- Reactant for Suzuki-Miyaura cross-coupling reaction
Vzorec:
C₃H₆BClO₂ Bod tání: 110…113 °C (lit.) |
MDL Number:
MFCD01074546 CAS číslo: 215951-86-3 |
Výsledky specifikačního testu
Colour | White to faint yellow |
Form | Powder or crystal or chunk |
Proton NMR spectrum | Conforms to structure |
Assay | ≥95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...