Tisk…

Hyaluronic acid sodium salt (from Streptococcus equi), Sigma-Aldrich®

Dodavatel: Merck

53747-10G 53747-1G
SIAL53747-10GEA 18200 CZK
SIAL53747-10G SIAL53747-1G
Hyaluronic acid sodium salt (from Streptococcus equi), Sigma-Aldrich®
Hyaluronic acid sodium salt

Hyaluronic acid sodium salt is isolated from Streptococcus equi. Used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.


mol wt ~1.5-1.8 x 10E6 Da
impurities ≤1% protein
solubility H2O: 5 mg/mL, clear, colorless
storage temp. −20°C

Vzorec: (C₁₄H₂₁NaNO₁₁)n
Storage Temperature: Freezer
MDL Number: MFCD00875848
CAS číslo: 9067-32-7

Objednat

Výsledky specifikačního testu

Impurities (protein) ≤1%
Solubility in H₂O 5 mg/ml, clear colourless
Storage temperature −20 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR