Tisk…

2-Phenoxyethyl acrylate ≥82%, Sigma-Aldrich®

Dodavatel: Merck
Varování


M-Clarity™ quality level = MQ100

408336-1L 408336-250ML
SIAL408336-1LEA 15220 CZK
SIAL408336-1L SIAL408336-250ML
2-Phenoxyethyl acrylate ≥82%, Sigma-Aldrich®
2-Phenoxyethyl acrylate

Ethylene glycol phenyl ether acrylate (PA) is a monofunctional acrylate. It contains 75 - 125 ppm hydroquinone as inhibitor, 0 - 120 ppm hydroquinone monomethyl as inhibitor.


Ethylene glycol phenyl ether acrylate (PA) monomer may be used to fabricate polymerisation induced grating pattern on photopolymer films.

Vzorec: C₁₁H₁₂O₃
Bod varu: 84 °C (0,2 mmHg) (l)
Hustota: 1,104 g/ml (25 °C)
Bod vzplanutí: 113 °C
MDL Number: MFCD00053696
CAS číslo: 48145-04-6
UN: 3082
ADR: 9,III

Objednat

Výsledky specifikačního testu

Colour Colourless
Form Liquid
Infrared spectrum Conforms to structure
Purity (GC) ≥82,0%
Hydroquinone as stabilizer 75 – 125 ppm
MEHQ as inhibitor ≤120 ppm
Refractive index n20/D 1,518 (l)
Density (25 °C) 1,104 g/ml

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR