Salmeterol xinafoate
Dodavatel: Thermo Scientific
Synonyms:
Salmeterol 1-hydroxy-2-naphthoate
462175000
462171000
ACRO462175000EA
14970
CZK
ACRO462175000
ACRO462171000
Salmeterol xinafoate
Salmeterol xinafoate
Vzorec:
C₂₅H₃₇NO₄·C₁₁H₈O₃ MW: 603,76 g/mol Bod tání: 126…128 °C Storage Temperature: Freezer |
MDL Number:
MFCD00897708 CAS číslo: 94749-08-3 |
Výsledky specifikačního testu
Appearance | White to almost white Powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...